Learn More
Electron Microscopy Sciences Autoclavable Instrument Tray 1, 102 mm x 178 mm, 54 mm depth
SDP
Catalog No. 5026736
Encompass_Preferred
Natural polypropylene. Steam autoclavable at 250 degree F (121 degree C) Smooth with rounded corners. They are easily cleaned and sterilized. The cover protects the instruments from contamination.
Dimensions: 102 mm x 178 mm
Depth: 54 mm
Catalog No. 50-267-36 Supplier Electron Microscopy Sciences Supplier No. 6331201
Natural polypropylene. Steam autoclavable at 250 degree F (121 degree C) Smooth with rounded corners. They are easily cleaned and sterilized. The cover protects the instruments from contamination.
Dimensions: 102 mm x 178 mm
Depth: 54 mm