Learn More
Sklar Instruments Instrument Tray, Sklar, Solid, Stainless Steel. The overall size of this tray is 10-3/8 inches x 6-3/8 inches x 2-1/2 inches. EA/1.
SDP
Catalog No. 501437114 Shop All Sklar Instruments Products
Encompass_Preferred
Instrument Tray, Sklar, Solid, Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this tray is 10-3/8 inches x 6-3/8 inches x 2-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7114 Supplier Sklar Instruments Supplier No. 101742
Instrument Tray, Sklar, Solid, Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this tray is 10-3/8 inches x 6-3/8 inches x 2-1/2 inches.