Learn More
Sklar Instruments Instrument Tray, Sklar, Solid, Stainless Steel. The overall size of this tray is 12-3/4 inches x 10-1/2 inches x 4 inches. EA/1.
SDP
Catalog No. 501437116 Shop All Sklar Instruments Products
Encompass_Preferred
Instrument Tray, Sklar, Solid, Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this tray is 12-3/4 inches x 10-1/2 inches x 4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7116 Supplier Sklar Instruments Supplier No. 101747
Instrument Tray, Sklar, Solid, Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this tray is 12-3/4 inches x 10-1/2 inches x 4 inches.