Learn More
Electron Microscopy Sciences Instrument Tray 310 mm x 427 mm, 73 mm depth
SDP
Catalog No. 5026733
Encompass_Preferred
These deep polypropylene instrument trays keep fluids from splashing. The smooth surface and rounded internal corners allow for easy cleaning. It is lightweight and noiseless as compared to heavy metal trays.
Dimensions: 310 mm x 427 mm
Depth: 73 mm
Catalog No. 50-267-33 Supplier Electron Microscopy Sciences Supplier No. 6331002
These deep polypropylene instrument trays keep fluids from splashing. The smooth surface and rounded internal corners allow for easy cleaning. It is lightweight and noiseless as compared to heavy metal trays.
Dimensions: 310 mm x 427 mm
Depth: 73 mm