Learn More
Electron Microscopy Sciences Autoclavable Instrument Tray 2, 121 mm x 229 mm, 83 mm depth
SDP
Catalog No. 5026734
Encompass_Preferred
Natural polypropylene. Steam autoclavable at 250 degree F (121 degree C) Smooth with rounded corners. They are easily cleaned and sterilized. The cover protects the instruments from contamination.
Dimensions: 121 mm x 229 mm
Depth: 83 mm
Catalog No. 50-267-34 Supplier Electron Microscopy Sciences Supplier No. 6331202
Natural polypropylene. Steam autoclavable at 250 degree F (121 degree C) Smooth with rounded corners. They are easily cleaned and sterilized. The cover protects the instruments from contamination.
Dimensions: 121 mm x 229 mm
Depth: 83 mm