Learn More
Electron Microscopy Sciences Autoclavable Instrument Tray 3, 152 mm x 279 mm, 127 mm depth
SDP
Catalog No. 5026735
Encompass_Preferred
Natural polypropylene. Steam autoclavable at 250 degree F (121 degree C) Smooth with rounded corners. They are easily cleaned and sterilized. The cover protects the instruments from contamination.
Dimensions: 152 mm x 279 mm
Depth: 127 mm
Catalog No. 50-267-35 Supplier Electron Microscopy Sciences Supplier No. 6331203
Natural polypropylene. Steam autoclavable at 250 degree F (121 degree C) Smooth with rounded corners. They are easily cleaned and sterilized. The cover protects the instruments from contamination.
Dimensions: 152 mm x 279 mm
Depth: 127 mm