Learn More
Roboz Surgical INSTRUMENT TRAY LID 12.25X7.75
SDP
Catalog No. NC9564144
Encompass
Instrument Tray Lid; 12.25 X 7.75 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles And Siz
Catalog No. NC9564144 Supplier Roboz Surgical Supplier No. RT1402L
Instrument Tray Lid; 12.25 X 7.75 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles And Siz