Learn More
MSC Liberty Safety Work Gloves, Pigskin Leather, Red Fleece Lined, Large, 1/Dozen
Catalog No. 5000684950
Encompass
These insulated driver work gloves are constructed from durable grain pigskin leather and feature a warm red fleece lining for cold-weather comfort. Designed for reliability in general-purpose and industrial applications, they incorporate ergonomic features to ensure a secure fit and improved dexterity.
  • Constructed from standard grain pigskin leather
  • Features red fleece lining for insulation
  • Keystone thumb design for improved mobility and comfort
  • Shirred elastic back provides a secure fit
  • Color-coded fabric hemmed cuff for easy size identification
  • Available in size Large
Catalog No. 50-006-84950 Supplier MSC Supplier No. 00127696
These insulated driver work gloves are constructed from durable grain pigskin leather and feature a warm red fleece lining for cold-weather comfort. Designed for reliability in general-purpose and industrial applications, they incorporate ergonomic features to ensure a secure fit and improved dexterity.
  • Constructed from standard grain pigskin leather
  • Features red fleece lining for insulation
  • Keystone thumb design for improved mobility and comfort
  • Shirred elastic back provides a secure fit
  • Color-coded fabric hemmed cuff for easy size identification
  • Available in size Large