Learn More
Saf T Pak Inc Saf-T-Pak STP-320 - Category B Frozen/Ambient Insulated Shipping System (UN 3373), 4/Case
SDP
Catalog No. nc9317515 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
The STP-320 is an Insulated Category B Shipping System designed for the shipment of temperature controlled shipments using phase change materials such as the SAF-T-TEMP line as well as Dry ice as an energy source. Ideal for long distance frozen shipments. Outer Box - OD 15.375 x 13.625 x 12.75”, Pre-printed with Biological Substance, Category B, UN 3373 and Exempt Human/Animal Specimen Markings, Orientation ArrowsInner Box - OD 6.5 x 6.5 x 8”Insulated Chest - ID 12 x 10.25 x 8”
Catalog No. NC9317515 Supplier Saf T Pak Inc Supplier No. STP320
The STP-320 is an Insulated Category B Shipping System designed for the shipment of temperature controlled shipments using phase change materials such as the SAF-T-TEMP line as well as Dry ice as an energy source. Ideal for long distance frozen shipments. Outer Box - OD 15.375 x 13.625 x 12.75”, Pre-printed with Biological Substance, Category B, UN 3373 and Exempt Human/Animal Specimen Markings, Orientation ArrowsInner Box - OD 6.5 x 6.5 x 8”Insulated Chest - ID 12 x 10.25 x 8”