Learn More
Ampco Insulated Adjustable Wrench
SDP
Catalog No. 19174124 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
7 -3/4 in.
Catalog No. Length (English)
19-174-124 7 -3/4 in.
1 options
Catalog No. 19-174-124 Supplier Ampco Supplier No. IW142R
Add to Cart
Add to Cart
Insulated, non-sparking, and corrosion-resistant sock wrench.
  • 1000 V Rated.
  • Please see image for additional information regarding measurements.
  • Size (a): 3/8 in.
  • Length: 7 and 3/4 in.
Specifications
Product Type Insulated Adjustable Wrench
Length (English) 7 -3/4 in.
WARNING: Cancer - www.P65Warnings.ca.gov