Learn More
Cole-Parmer ThermoSafe 390 Dry Ice Storage Insulated Field Carrier, polyethylene, 1 cu ft
Catalog No. NC0653492 Shop All Cole-Parmer Products
Encompass
Insulated field carrier has efficient, 2-1/4" thick polyurethane foam insulation core and rugged, double-wall polyethylene construction. Includes carrying handle, hinged lid, and adjustable latches. Inside dimensions are 16-5/8"L x 10-1/4"W x 10-3/4"H.
Catalog No. NC0653492 Supplier Cole-Parmer Supplier No. 0372020
Insulated field carrier has efficient, 2-1/4" thick polyurethane foam insulation core and rugged, double-wall polyethylene construction. Includes carrying handle, hinged lid, and adjustable latches. Inside dimensions are 16-5/8"L x 10-1/4"W x 10-3/4"H.