Learn More
MSC MCR Safety Driver Gloves, Pigskin Leather, Fleece Lined, Slip-On, 1 ea.
Catalog No. 5000687675
Encompass
These driver-style work gloves are designed for general-purpose applications that require abrasion resistance and durability. Featuring a grain pigskin leather palm and a comfortable fleece lining, these gloves provide both protection and insulation, making them suitable for use in wet or cold environments.
  • Grain pigskin leather construction
  • Red fleece lining for insulation in cold conditions
  • Keystone thumb for increased mobility
  • Slip-on cuff with elastic/shirred closure for a secure fit
  • Gunn cut design for enhanced comfort
  • Suitable for carpentry, farming, landscaping, and cold storage applications
Catalog No. 50-006-87675 Supplier MSC Supplier No. 14157192
These driver-style work gloves are designed for general-purpose applications that require abrasion resistance and durability. Featuring a grain pigskin leather palm and a comfortable fleece lining, these gloves provide both protection and insulation, making them suitable for use in wet or cold environments.
  • Grain pigskin leather construction
  • Red fleece lining for insulation in cold conditions
  • Keystone thumb for increased mobility
  • Slip-on cuff with elastic/shirred closure for a secure fit
  • Gunn cut design for enhanced comfort
  • Suitable for carpentry, farming, landscaping, and cold storage applications