Learn More
Safe Guard Technologies INSULATED MAILER W/CRTN 30/CS
Catalog No. NC0291434
Encompass
Mailer Boxes: 8x6x2 Safe-Guard insulated containers for hard-to-ship preparations and laboratory handling of frozen biologicals, biopsy materials, frozen tissue, serums, animal parts, vaccines, human organs, pathology specimens, blood and virus samples and other temperature sensitive materials presenting laboratory shipping problems.
Catalog No. NC0291434 Supplier Safe Guard Technologies Supplier No. SG862
Mailer Boxes: 8x6x2 Safe-Guard insulated containers for hard-to-ship preparations and laboratory handling of frozen biologicals, biopsy materials, frozen tissue, serums, animal parts, vaccines, human organs, pathology specimens, blood and virus samples and other temperature sensitive materials presenting laboratory shipping problems.