Learn More
Ampco Insulated Pliers
SDP
Catalog No. 19174106 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
10 -1/4 in.
8 -3/16 in.
8 -3/4 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-174-106 10 -1/4 in.
19-174-098 8 -3/16 in.
19-174-099 8 -3/4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-174-106 Supplier Ampco Supplier No. IP39
Add to Cart
Add to Cart
Insulated, non-sparking, non-magnetic, and corrosion-resistant pliers.
  • 1000 V rated.
  • Made in the USA.
Specifications
Product Type Insulated Pliers
Length (English) 10 -1/4 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov