Learn More
Andwin Scientific Category B & Exempt Shipping System, Insulated
SDP
Catalog No. NC9563908
Encompass_Preferred
System Components: Outer Box OD 15 3/8 x 13 5/8 x 12 3/4" pre-printed with Biological Substance, Category B, UN 3373 and Exempt Human/Animal Specimen Markings. Insulated Chest ID 11 5/8 x 10 x 8", STP-111 Inner Box OD 6 1/2 x 6 1/2 x 8", STP-152 250 mL, Absorbent Strip (2 ea.), STP-609 6 x 12" Saf-T-Rap Bubble Wrap (2 ea.), STP-710 Disposable 2-Part Secondary Pressure Vessel, Medium (2 ea.), STP-803 Class 9 Hazard Label (1 ea.), STP-804 Dry Ice Marking (1 ea.).
Catalog No. NC9563908 Supplier Andwin Scientific Supplier No. STP320
System Components: Outer Box OD 15 3/8 x 13 5/8 x 12 3/4" pre-printed with Biological Substance, Category B, UN 3373 and Exempt Human/Animal Specimen Markings. Insulated Chest ID 11 5/8 x 10 x 8", STP-111 Inner Box OD 6 1/2 x 6 1/2 x 8", STP-152 250 mL, Absorbent Strip (2 ea.), STP-609 6 x 12" Saf-T-Rap Bubble Wrap (2 ea.), STP-710 Disposable 2-Part Secondary Pressure Vessel, Medium (2 ea.), STP-803 Class 9 Hazard Label (1 ea.), STP-804 Dry Ice Marking (1 ea.).