Learn More
MSC The Packaging Wholesalers Insulated Shipping Container, 8x6x12 in, 1 ea
Catalog No. 501920724
Encompass
These insulated shipping containers are designed to maintain the insulation and integrity of temperature-sensitive products. Each kit includes a pre-assembled 200#/ECT-32 white corrugated carton with a seamless molded EPS foam container and a tight-fitting lid.
  • Maintains ideal temperature for contents during transit
  • Lightweight design helps reduce shipping costs and facilitates easy transport
  • Durable foam construction protects contents against damage
  • Cartons are printed with 'Rush,' 'Perishable,' and 'This Side Up' shipping instructions
  • Materials are reusable and recyclable for an eco-friendly shipping solution
Catalog No. 50-192-0724 Supplier MSC Supplier No. 39524202
These insulated shipping containers are designed to maintain the insulation and integrity of temperature-sensitive products. Each kit includes a pre-assembled 200#/ECT-32 white corrugated carton with a seamless molded EPS foam container and a tight-fitting lid.
  • Maintains ideal temperature for contents during transit
  • Lightweight design helps reduce shipping costs and facilitates easy transport
  • Durable foam construction protects contents against damage
  • Cartons are printed with 'Rush,' 'Perishable,' and 'This Side Up' shipping instructions
  • Materials are reusable and recyclable for an eco-friendly shipping solution