Learn More
TA Instruments Insulated Solvent Trap, Discovery Core Rheometer, HR X0
Catalog No. 5000580787 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Insulated Solvent Trap for the Discovery Core Rheometer prevents evaporation at the sample edge in cone and plate or parallel plate configurations. When used with a solvent trap-compatible geometry, the solvent trap flange extends into the geometry solvent well which can be filled with liquid to create a seal around the sample, without physical contact with the measuring tool. The split-cover design fits closely around the Peltier plate. A conductive interior and insulated exterior maximize temperature uniformity around the sample.
Catalog No. 50-005-80787 Supplier TA Instruments Supplier No. 536500.901
Waters TA Instruments Insulated Solvent Trap for the Discovery Core Rheometer prevents evaporation at the sample edge in cone and plate or parallel plate configurations. When used with a solvent trap-compatible geometry, the solvent trap flange extends into the geometry solvent well which can be filled with liquid to create a seal around the sample, without physical contact with the measuring tool. The split-cover design fits closely around the Peltier plate. A conductive interior and insulated exterior maximize temperature uniformity around the sample.