Learn More
Ampco Insulated Standard Screwdriver
SDP
Catalog No. 19174110 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
10 -1/4 in.
Catalog No. Length (English)
19-174-110 10 -1/4 in.
1 options
Catalog No. 19-174-110 Supplier Ampco Supplier No. IS49
Add to Cart
Add to Cart
Non-sparking.
  • See image for additional information.
  • Tip size (a): 5/16 in.
  • Insulated tip length (b): 6 in.
  • Overall length: 10 and 1/4 in.
Specifications
Product Type Insulated Screwdriver
Length (English) 10 -1/4 in.
WARNING: Cancer - www.P65Warnings.ca.gov