Learn More
Grainger Salisbury Electrical Insulating Blanket, Class IV, Synthetic, 3 ft x 3 ft, Orange
Catalog No. NC2865693
Encompass_Preferred
This synthetic electrical insulating blanket is designed for live-front work on high-voltage equipment. Rated as Class IV, it provides reliable insulation in demanding electrical applications and complies with ASTM F2320-18 standards.
  • Class IV protection for high-voltage environments
  • Maximum use voltage of 36,000 V AC and 54,000 V DC
  • ASTM F2320-18 compliant for safety and performance
  • Features a durable, ozone-resistant synthetic construction
  • Includes integrated eyelets for secure installation
  • Equipped with a date stamp for tracking and maintenance
  • Dimensions of 91.4 cm x 91.4 cm (3 ft. x 3 ft.)
  • AC proof test voltage of 40,000 V rms and DC proof test voltage of 70,000 V avg
Catalog No. NC2865693 Supplier Grainger Supplier No. 5ZV86
This synthetic electrical insulating blanket is designed for live-front work on high-voltage equipment. Rated as Class IV, it provides reliable insulation in demanding electrical applications and complies with ASTM F2320-18 standards.
  • Class IV protection for high-voltage environments
  • Maximum use voltage of 36,000 V AC and 54,000 V DC
  • ASTM F2320-18 compliant for safety and performance
  • Features a durable, ozone-resistant synthetic construction
  • Includes integrated eyelets for secure installation
  • Equipped with a date stamp for tracking and maintenance
  • Dimensions of 91.4 cm x 91.4 cm (3 ft. x 3 ft.)
  • AC proof test voltage of 40,000 V rms and DC proof test voltage of 70,000 V avg