Learn More
Cole-Parmer Powerblanket PBICE55IP Insulating Cooling Blanket with Ice Packs; 55 Gallon
Catalog No. NC1979228 Shop All Cole-Parmer Products
Encompass
These insulating cooling drum and pail blankets are an easy and cost-effective way to cool large vessels containing temperature sensitive materials. Simply place the ice packs in the pockets on the inside surface of the blanket and place the blanket on to the drum. Modify the number of ice packs used to regulate temperature. Built-in straps securely cinch blanket to the outside of the drum for optimum performance and enhanced portability.
Catalog No. NC1979228 Supplier Cole-Parmer Supplier No. 4202528
These insulating cooling drum and pail blankets are an easy and cost-effective way to cool large vessels containing temperature sensitive materials. Simply place the ice packs in the pockets on the inside surface of the blanket and place the blanket on to the drum. Modify the number of ice packs used to regulate temperature. Built-in straps securely cinch blanket to the outside of the drum for optimum performance and enhanced portability.