Learn More
Chemglass Life Sciences 10L Insulating Jacket for Jacketed Cylindrical Reactor (fits Chemglass 10 L jacketed reactors)
SDP
Catalog No. NC3355229
Encompass_Preferred
The Chemglass 10L insulating jacket is a removable thermal jacket designed to fit 10 L jacketed cylindrical reactors. It insulates the reactor to help maintain process temperatures during heating or cooling and is constructed for laboratory durability with fiberglass and silicone-coated materials. CSA approved.
  • Provides thermal insulation for 10 L jacketed reactors
  • Constructed with heavy fiberglass fabric interior
  • Outer layer of silicone-coated fiberglass for chemical resistance
  • Slit and lacing closure to accommodate bottom drain valves
  • Removable and reusable for repeated lab procedures
  • Certified to CSA safety standards
Catalog No. NC3355229 Supplier Chemglass Life Sciences Supplier No. CG10011110
The Chemglass 10L insulating jacket is a removable thermal jacket designed to fit 10 L jacketed cylindrical reactors. It insulates the reactor to help maintain process temperatures during heating or cooling and is constructed for laboratory durability with fiberglass and silicone-coated materials. CSA approved.
  • Provides thermal insulation for 10 L jacketed reactors
  • Constructed with heavy fiberglass fabric interior
  • Outer layer of silicone-coated fiberglass for chemical resistance
  • Slit and lacing closure to accommodate bottom drain valves
  • Removable and reusable for repeated lab procedures
  • Certified to CSA safety standards