Learn More
Grainger Johns Manville High Temperature Insulation Board, Calcium Silicate, 1700 °F, 8 ft x 4 ft, 1 ea
Catalog No. 502779852
Encompass_Preferred
This calcium silicate board is a rigid, high-temperature insulation solution designed for industrial applications requiring exceptional structural strength. It offers high-compression resistance and effective thermal management across a wide operating temperature range.
  • Temperature range of 0°F to 1,700°F
  • Calcium silicate material composition
  • 1-1/2 in. thickness
  • Unfaced design
  • Minimal moisture and water vapor absorption
  • Extra extra strong construction
  • Complies with ASTM C656, C795, E119, E136, E72, E84, NFPA251, UL 1479, UL 1709, and UL 263 standards
Catalog No. 50-277-9852 Supplier Grainger Supplier No. 19NE58
This calcium silicate board is a rigid, high-temperature insulation solution designed for industrial applications requiring exceptional structural strength. It offers high-compression resistance and effective thermal management across a wide operating temperature range.
  • Temperature range of 0°F to 1,700°F
  • Calcium silicate material composition
  • 1-1/2 in. thickness
  • Unfaced design
  • Minimal moisture and water vapor absorption
  • Extra extra strong construction
  • Complies with ASTM C656, C795, E119, E136, E72, E84, NFPA251, UL 1479, UL 1709, and UL 263 standards