Learn More
Grainger Johns Manville Thermo-1200 Calcium Silicate Insulation, Water Resistant, Type I, 1,200°F
Catalog No. 502783187
Encompass_Preferred
This product is a Type I, high-temperature calcium silicate insulation designed for systems with operating temperatures ranging from ambient up to 1200°F (650°C). Formulated with a proprietary XOX Corrosion Inhibitor®, it provides an effective barrier to help mitigate corrosion under insulation (CUI). Its water-resistant properties allow for installation flexibility by enabling water to bead on the surface rather than penetrating the substrate during brief exposure to weather.
  • Meets or exceeds physical and thermal property requirements of ASTM C533, Type 1
  • Non-combustible and asbestos-free composition
  • Engineered to resist water absorption during heavy rainfall events
  • Helps protect against corrosion under insulation
  • Available in various configurations including pipe, block, and curved segments
Catalog No. 50-278-3187 Supplier Grainger Supplier No. 19NE60
This product is a Type I, high-temperature calcium silicate insulation designed for systems with operating temperatures ranging from ambient up to 1200°F (650°C). Formulated with a proprietary XOX Corrosion Inhibitor®, it provides an effective barrier to help mitigate corrosion under insulation (CUI). Its water-resistant properties allow for installation flexibility by enabling water to bead on the surface rather than penetrating the substrate during brief exposure to weather.
  • Meets or exceeds physical and thermal property requirements of ASTM C533, Type 1
  • Non-combustible and asbestos-free composition
  • Engineered to resist water absorption during heavy rainfall events
  • Helps protect against corrosion under insulation
  • Available in various configurations including pipe, block, and curved segments