Learn More
Grainger Johns Manville Thermo-1200 Calcium Silicate Board Insulation, 1.5 in Thick
Catalog No. 502783189
Encompass_Preferred
Thermo-1200 is a water-resistant, Type I calcium silicate block insulation engineered for demanding high-temperature industrial applications. Designed for use at service temperatures up to 1200°F (650°C), this inorganic, non-combustible material provides exceptional thermal efficiency and structural integrity. It is formulated with XOX Corrosion Inhibitor® to help protect pipe and vessel surfaces from corrosion under insulation.
  • Service temperature up to 1200°F (650°C)
  • Contains XOX Corrosion Inhibitor® to protect against surface corrosion
  • Water-resistant design withstands heavy rainfall
  • Non-combustible composition
  • Meets ASTM C533, Type I requirements
  • Asbestos, lead, and mercury-free
  • Compressive strength of 100 psi
  • Meets UL 1709 rapid rise fire test requirements
Catalog No. 50-278-3189 Supplier Grainger Supplier No. 19NE72
Thermo-1200 is a water-resistant, Type I calcium silicate block insulation engineered for demanding high-temperature industrial applications. Designed for use at service temperatures up to 1200°F (650°C), this inorganic, non-combustible material provides exceptional thermal efficiency and structural integrity. It is formulated with XOX Corrosion Inhibitor® to help protect pipe and vessel surfaces from corrosion under insulation.
  • Service temperature up to 1200°F (650°C)
  • Contains XOX Corrosion Inhibitor® to protect against surface corrosion
  • Water-resistant design withstands heavy rainfall
  • Non-combustible composition
  • Meets ASTM C533, Type I requirements
  • Asbestos, lead, and mercury-free
  • Compressive strength of 100 psi
  • Meets UL 1709 rapid rise fire test requirements