Learn More
Grainger Johns Manville Calcium Silicate Insulation, 1-1/2 In. Thick, 12 In. X 12 In., 1 Ea.
Catalog No. 502779853
Encompass_Preferred
This high-temperature, Type I calcium silicate insulation is designed for industrial applications requiring thermal management at temperatures up to 1200°F (650°C). Its cementitious construction provides durability and physical integrity in demanding environments.
  • Engineered with an integral corrosion inhibitor to protect against corrosion under insulation (CUI)
  • High compressive strength (>100 psi) for mechanical abuse resistance
  • Non-combustible and free of asbestos, lead, and mercury
  • Water-resistant design withstands heavy rainfall without excessive absorption
  • Maintains structural integrity at elevated temperatures where organic binders fail
  • Complies with ASTM C533, Type I and various industrial fire safety standards
Catalog No. 50-277-9853 Supplier Grainger Supplier No. 19NE71
This high-temperature, Type I calcium silicate insulation is designed for industrial applications requiring thermal management at temperatures up to 1200°F (650°C). Its cementitious construction provides durability and physical integrity in demanding environments.
  • Engineered with an integral corrosion inhibitor to protect against corrosion under insulation (CUI)
  • High compressive strength (>100 psi) for mechanical abuse resistance
  • Non-combustible and free of asbestos, lead, and mercury
  • Water-resistant design withstands heavy rainfall without excessive absorption
  • Maintains structural integrity at elevated temperatures where organic binders fail
  • Complies with ASTM C533, Type I and various industrial fire safety standards