Learn More
Grainger Johns Manville Calcium Silicate High Temperature Insulation Board, 4 ft x 4 ft, 0.5 in Thick, 1 ea
Catalog No. 502783341
Encompass_Preferred
This calcium silicate board is a high-temperature insulation material designed for extreme environments. It is a rigid, lightweight board featuring high compressive strength and minimal moisture absorption for effective thermal performance. The material is suitable for demanding applications requiring consistent insulation properties up to 1,700 °F.
  • Temperature range of 0 °F to 1,700 °F
  • Rigid calcium silicate board construction
  • Minimal water vapor absorption
  • White, unfaced surface
  • Meets ASTM C656, C795, E119, E136, E72, E84, NFPA251, UL 1479, UL 1709 and UL 263 standards
Catalog No. 50-278-3341 Supplier Grainger Supplier No. 19NE61
This calcium silicate board is a high-temperature insulation material designed for extreme environments. It is a rigid, lightweight board featuring high compressive strength and minimal moisture absorption for effective thermal performance. The material is suitable for demanding applications requiring consistent insulation properties up to 1,700 °F.
  • Temperature range of 0 °F to 1,700 °F
  • Rigid calcium silicate board construction
  • Minimal water vapor absorption
  • White, unfaced surface
  • Meets ASTM C656, C795, E119, E136, E72, E84, NFPA251, UL 1479, UL 1709 and UL 263 standards