Learn More
Grainger Johns Manville Calcium Silicate Block Insulation, 1700°F, 1/2 in. Thick
Catalog No. 502779847
Encompass_Preferred
This high-temperature block insulation is an inorganic, non-combustible material composed primarily of hydrous calcium silicate. Designed for high-performance thermal management, it is suitable for systems operating at temperatures up to 1700°F (927°C). It meets or exceeds the physical and thermal requirements of ASTM C533, Type II, and provides reliable insulation for curved vessels, large diameter pipes, boilers, ducts, and industrial flues.
  • Provides excellent resistance to damage, enhancing the life of the system.
  • Tested and compliant with UL 1709 (Section 9) standards for rapid rise fire protection.
  • Features a non-combustible formulation meeting ASTM E136 requirements.
  • Passes rigorous corrosion testing for austenitic stainless steel.
  • Essentially dust-free and contains no asbestos.
Catalog No. 50-277-9847 Supplier Grainger Supplier No. 19NE46
This high-temperature block insulation is an inorganic, non-combustible material composed primarily of hydrous calcium silicate. Designed for high-performance thermal management, it is suitable for systems operating at temperatures up to 1700°F (927°C). It meets or exceeds the physical and thermal requirements of ASTM C533, Type II, and provides reliable insulation for curved vessels, large diameter pipes, boilers, ducts, and industrial flues.
  • Provides excellent resistance to damage, enhancing the life of the system.
  • Tested and compliant with UL 1709 (Section 9) standards for rapid rise fire protection.
  • Features a non-combustible formulation meeting ASTM E136 requirements.
  • Passes rigorous corrosion testing for austenitic stainless steel.
  • Essentially dust-free and contains no asbestos.