Learn More
Grainger Johns Manville Thermo-1200 Pipe and Board Insulation, Calcium Silicate, Water Resistant, 12 in x 12 in
Catalog No. 502779813
Encompass_Preferred
This water-resistant, Type I calcium silicate insulation is engineered for industrial pipe, block, and curved segment applications. Formulated with an inorganic binder and a corrosion inhibitor, it provides thermal protection for systems operating at temperatures up to 1200°F (649°C). This non-combustible material is designed to maintain structural integrity and minimize the potential for corrosion under insulation.
  • Service temperature up to 1200°F (649°C)
  • Features corrosion inhibitor to minimize surface corrosion
  • Non-combustible material compliant with ASTM C533, Type 1
  • Water-resistant design withstands 1.25 in./hr. of rainfall for up to 20 minutes
  • Asbestos, lead, and mercury-free composition
  • Compressive strength greater than 100 psi
  • Estimated lifespan of 25 years or more
Catalog No. 50-277-9813 Supplier Grainger Supplier No. 19NE67
This water-resistant, Type I calcium silicate insulation is engineered for industrial pipe, block, and curved segment applications. Formulated with an inorganic binder and a corrosion inhibitor, it provides thermal protection for systems operating at temperatures up to 1200°F (649°C). This non-combustible material is designed to maintain structural integrity and minimize the potential for corrosion under insulation.
  • Service temperature up to 1200°F (649°C)
  • Features corrosion inhibitor to minimize surface corrosion
  • Non-combustible material compliant with ASTM C533, Type 1
  • Water-resistant design withstands 1.25 in./hr. of rainfall for up to 20 minutes
  • Asbestos, lead, and mercury-free composition
  • Compressive strength greater than 100 psi
  • Estimated lifespan of 25 years or more