Learn More
Grainger INSULATION CALCIUM SILICATE 1X
Catalog No. 502783149
Encompass_Preferred
This is a water-resistant, Type I calcium silicate block insulation designed for high-temperature applications up to 1200°F (650°C). It incorporates a specialized corrosion inhibiting formula to help protect against corrosion under insulation in environments such as chemical processing, power generation, and petroleum refining.
  • Water resistant and non-combustible material
  • Contains corrosion inhibitor to protect against corrosion under insulation
  • Asbestos, lead, and mercury-free formulation
  • Suitable for service temperatures up to 1200°F (650°C)
  • Features excellent compressive and flexural strength
  • Meets or exceeds ASTM C533, Type 1 standards
Catalog No. 50-278-3149 Supplier Grainger Supplier No. 19NE54
This is a water-resistant, Type I calcium silicate block insulation designed for high-temperature applications up to 1200°F (650°C). It incorporates a specialized corrosion inhibiting formula to help protect against corrosion under insulation in environments such as chemical processing, power generation, and petroleum refining.
  • Water resistant and non-combustible material
  • Contains corrosion inhibitor to protect against corrosion under insulation
  • Asbestos, lead, and mercury-free formulation
  • Suitable for service temperatures up to 1200°F (650°C)
  • Features excellent compressive and flexural strength
  • Meets or exceeds ASTM C533, Type 1 standards