Learn More
Grainger Johns Manville Thermo-1200 Calcium Silicate Insulation Board, 1 in Thick, 4 ft x 8 ft
Catalog No. 502783188
Encompass_Preferred
This high temperature calcium silicate insulation board is a cementitious, water-resistant material designed for demanding industrial applications. It is engineered with an inorganic binder to maintain physical integrity and shape even in extreme heat environments, providing reliable thermal performance for service temperatures up to 1200°F.
  • Service temperature up to 1200°F
  • Features XOX Corrosion Inhibitor® to help protect against corrosion under insulation
  • Exceptional compressive strength of 100 psi for resistance to mechanical abuse
  • Water resistant to withstand heavy rainfall during installation
  • Inorganic binder maintains physical integrity past 450°F
  • Asbestos, lead, and mercury-free composition
  • Meets ASTM C533, Type 1 specifications
  • Potential insulation lifespan of up to 25 years or more when properly maintained
Catalog No. 50-278-3188 Supplier Grainger Supplier No. 19NE70
This high temperature calcium silicate insulation board is a cementitious, water-resistant material designed for demanding industrial applications. It is engineered with an inorganic binder to maintain physical integrity and shape even in extreme heat environments, providing reliable thermal performance for service temperatures up to 1200°F.
  • Service temperature up to 1200°F
  • Features XOX Corrosion Inhibitor® to help protect against corrosion under insulation
  • Exceptional compressive strength of 100 psi for resistance to mechanical abuse
  • Water resistant to withstand heavy rainfall during installation
  • Inorganic binder maintains physical integrity past 450°F
  • Asbestos, lead, and mercury-free composition
  • Meets ASTM C533, Type 1 specifications
  • Potential insulation lifespan of up to 25 years or more when properly maintained