Learn More
Grainger Johns Manville Calcium Silicate Insulation, Board, Type I, 1,700°F
Catalog No. 502780031
Encompass_Preferred
This is a water-resistant, Type I calcium silicate insulation board designed for high-temperature applications up to 1,200°F (650°C). Engineered with an inorganic binder, this cementitious insulation maintains its physical integrity even after organic binders typically burn off, providing robust performance in demanding industrial environments.
  • Features corrosion inhibitor to protect against corrosion under insulation (CUI).
  • Provides exceptional compressive strength (>100 psi) suitable for areas with high mechanical abuse.
  • Non-combustible and asbestos-free construction.
  • Engineered to withstand heavy rainfall for up to 20 minutes with minimal water absorption.
  • Offers a long service life of up to 25 years when properly maintained.
  • Complies with ASTM C533, Type I material specifications.
Catalog No. 50-278-0031 Supplier Grainger Supplier No. 19NE66
This is a water-resistant, Type I calcium silicate insulation board designed for high-temperature applications up to 1,200°F (650°C). Engineered with an inorganic binder, this cementitious insulation maintains its physical integrity even after organic binders typically burn off, providing robust performance in demanding industrial environments.
  • Features corrosion inhibitor to protect against corrosion under insulation (CUI).
  • Provides exceptional compressive strength (>100 psi) suitable for areas with high mechanical abuse.
  • Non-combustible and asbestos-free construction.
  • Engineered to withstand heavy rainfall for up to 20 minutes with minimal water absorption.
  • Offers a long service life of up to 25 years when properly maintained.
  • Complies with ASTM C533, Type I material specifications.