Learn More
Grainger Johns Manville Thermo-1200 Calcium Silicate Insulation, Board, 3/4 in.
Catalog No. 502779785
Encompass_Preferred
This is a type I calcium silicate pipe and block insulation engineered for high-temperature industrial applications. It features a proprietary corrosion inhibitor to protect against pipe corrosion and offers superior durability in demanding environments.
  • Equipped with a proprietary corrosion inhibitor to mitigate surface corrosion
  • Engineered to be water resistant, capable of withstanding heavy rainfall of 1 1/4 in. per hour for up to 20 min. with minimal moisture absorption
  • Non-combustible material with a flame spread and smoke developed index of 0
  • Certified as asbestos, lead, and mercury-free
  • Meets ASTM C533, Type I material specifications
  • Excellent thermal performance with a service temperature up to 1,200°F (649°C)
  • Passes rigorous industry tests including corrosion testing on austenitic stainless steel
Catalog No. 50-277-9785 Supplier Grainger Supplier No. 19NE63
This is a type I calcium silicate pipe and block insulation engineered for high-temperature industrial applications. It features a proprietary corrosion inhibitor to protect against pipe corrosion and offers superior durability in demanding environments.
  • Equipped with a proprietary corrosion inhibitor to mitigate surface corrosion
  • Engineered to be water resistant, capable of withstanding heavy rainfall of 1 1/4 in. per hour for up to 20 min. with minimal moisture absorption
  • Non-combustible material with a flame spread and smoke developed index of 0
  • Certified as asbestos, lead, and mercury-free
  • Meets ASTM C533, Type I material specifications
  • Excellent thermal performance with a service temperature up to 1,200°F (649°C)
  • Passes rigorous industry tests including corrosion testing on austenitic stainless steel