Learn More
Grainger Johns Manville Thermo-1200 Calcium Silicate Insulation, Water Resistant, 1200°F
Catalog No. 502779814
Encompass_Preferred
This Thermo-1200 high-temperature insulation is a water-resistant, Type I calcium silicate material engineered for demanding industrial thermal applications. It features an integrated corrosion inhibitor to protect against corrosion under insulation (CUI) and provides high compressive strength to withstand mechanical stress. The insulation is designed for long-term durability and is free of asbestos, lead, and mercury.
  • Service temperature up to 1200°F (maximum use 1700°F)
  • Water resistant to withstand heavy rainfall during installation
  • Includes XOX Corrosion Inhibitor to minimize CUI
  • Cementitious design with compressive strength of 100 psi
  • Non-combustible and inorganic construction for stability
  • Provides an expected lifespan of 25 years or more
  • Meets ASTM C533, Type 1 standards
Catalog No. 50-277-9814 Supplier Grainger Supplier No. 19NE64
This Thermo-1200 high-temperature insulation is a water-resistant, Type I calcium silicate material engineered for demanding industrial thermal applications. It features an integrated corrosion inhibitor to protect against corrosion under insulation (CUI) and provides high compressive strength to withstand mechanical stress. The insulation is designed for long-term durability and is free of asbestos, lead, and mercury.
  • Service temperature up to 1200°F (maximum use 1700°F)
  • Water resistant to withstand heavy rainfall during installation
  • Includes XOX Corrosion Inhibitor to minimize CUI
  • Cementitious design with compressive strength of 100 psi
  • Non-combustible and inorganic construction for stability
  • Provides an expected lifespan of 25 years or more
  • Meets ASTM C533, Type 1 standards