This Type I calcium silicate insulation is engineered for high-temperature applications reaching up to 1200°F (650°C). Featuring integral XOX Corrosion Inhibitor®, it is designed to protect against corrosion under insulation while maintaining structural integrity. Its cementitious composition is water-resistant, capable of withstanding heavy rainfall of 1¼ in. per hour for up to 20 minutes without exceeding 15% water absorption by weight.
- Water-resistant design minimizes moisture absorption to less than 15% by weight.
- Compressive strength of 100 psi supports use in environments prone to mechanical abuse.
- Inorganic binder maintains physical shape and integrity beyond 450°F.
- Non-combustible material compliant with ASTM standards.
- Designed for an insulation lifespan of up to 25 years with proper maintenance.