Learn More
Cole-Parmer Fluke 1577 Digital Insulation Multimeter
Supplier:  Cole-Parmer 2682810
Encompass
These meters are ideal for troubleshooting and conducting preventive maintenance on motors, switchgear, generators, and cables. Meters feature AC/DC voltage, DC mV, AC/DC mA, resistance and continuity measurement capabilities. Model 26828-20 provides capacitance, diode test, temperature, min/max, frequency (Hz), and filter for motor drive measurements. The large backlit LCD display is easy to read in dimly lit work environments and the auto power off function saves battery power.
Catalog No. 50-630-940