Learn More
Grainger Johns Manville Polyisocyanurate Insulation Board, FSK Facing, 24 in Width, 4 ft Length
Catalog No. 502778720
Encompass_Preferred
This polyisocyanurate foam insulation board is a closed-cell, high-performance material designed for thermal insulation in commercial and industrial HVAC, ductwork, and process equipment. It features FSK facing to provide a durable vapor barrier and enhanced rigidity.
  • Features a closed-cell polyisocyanurate foam core for high thermal resistance.
  • Includes FSK facing for durability and vapor barrier integrity.
  • Meets ASTM C 1289, Type II, Class 1, Grade 2 standards.
  • Provides service temperature range from -73°C to +121°C.
  • Compatible with diverse roofing systems including BUR, modified bitumen, and single-ply membranes.
  • Utilizes a pentane blowing agent with zero ozone depletion potential.
  • Tested for fire performance under UL 790, 263, and 1256 standards.
Catalog No. 50-277-8720 Supplier Grainger Supplier No. 19NE95
This polyisocyanurate foam insulation board is a closed-cell, high-performance material designed for thermal insulation in commercial and industrial HVAC, ductwork, and process equipment. It features FSK facing to provide a durable vapor barrier and enhanced rigidity.
  • Features a closed-cell polyisocyanurate foam core for high thermal resistance.
  • Includes FSK facing for durability and vapor barrier integrity.
  • Meets ASTM C 1289, Type II, Class 1, Grade 2 standards.
  • Provides service temperature range from -73°C to +121°C.
  • Compatible with diverse roofing systems including BUR, modified bitumen, and single-ply membranes.
  • Utilizes a pentane blowing agent with zero ozone depletion potential.
  • Tested for fire performance under UL 790, 263, and 1256 standards.