Learn More
Grainger Itw Polyisocyanurate High Temperature Insulation, Board, 24 in x 48 in x 1 1/2 in, 1 ea
Catalog No. 502782971
Encompass_Preferred
This polyisocyanurate (PIR) foam board provides high-temperature thermal insulation in a closed-cell structure. Designed for demanding thermal environments, it maintains performance across a wide operating temperature range.
  • Operating temperature range of -297°F to 297°F
  • Density of 2 lb/cu ft
  • R-value of 7.9
  • Water vapor permeability of 0.02 perm
  • Unfaced board form
  • Compliant with ASTM D1622, C518, D2126, E84, and E96 standards
Catalog No. 50-278-2971 Supplier Grainger Supplier No. 19NE87
This polyisocyanurate (PIR) foam board provides high-temperature thermal insulation in a closed-cell structure. Designed for demanding thermal environments, it maintains performance across a wide operating temperature range.
  • Operating temperature range of -297°F to 297°F
  • Density of 2 lb/cu ft
  • R-value of 7.9
  • Water vapor permeability of 0.02 perm
  • Unfaced board form
  • Compliant with ASTM D1622, C518, D2126, E84, and E96 standards