Learn More
Grainger ITW High Temperature Insulation, PIR Foam, 24 in. W, 48 in. L, 1-1/2 in. Thick
Catalog No. 502778967
Encompass_Preferred
These rigid insulation boards are constructed from polyisocyanurate (PIR) foam and are designed for high-temperature applications. Featuring FSK facing, they offer a lightweight yet durable solution for thermal insulation that is stronger than conventional blankets and wraps.
  • Provides an R-value of 7.9
  • Operates within a temperature range of -297°F to +297°F
  • Features high-compression strength and a closed foam cell type
  • Measures 24 in. width, 4 ft. length, and 1 1/2 in. thickness
  • Engineered with high moisture resistance at 0.02 perms
  • Meets ASTM D1622, C518, D2126, E84, and E96 standards
Catalog No. 50-277-8967 Supplier Grainger Supplier No. 19NE92
These rigid insulation boards are constructed from polyisocyanurate (PIR) foam and are designed for high-temperature applications. Featuring FSK facing, they offer a lightweight yet durable solution for thermal insulation that is stronger than conventional blankets and wraps.
  • Provides an R-value of 7.9
  • Operates within a temperature range of -297°F to +297°F
  • Features high-compression strength and a closed foam cell type
  • Measures 24 in. width, 4 ft. length, and 1 1/2 in. thickness
  • Engineered with high moisture resistance at 0.02 perms
  • Meets ASTM D1622, C518, D2126, E84, and E96 standards