Learn More
Grainger ITW High Temperature Insulation, PIR Foam, FSK Facing, 24 in W, 4 ft L, 2 in Thick, 1 ea
Catalog No. 502782449
Encompass_Preferred
These insulation boards are manufactured from polyisocyanurate (PIR) foam and feature FSK facing. Designed for high-performance thermal insulation, these boards are lightweight, rigid, and offer high-compression strength, making them a robust alternative to traditional insulation blankets and wraps. This insulation is suitable for applications requiring a temperature range of -297°F to +297°F.
  • Closed cell foam construction
  • Thermal resistance of 10.5 R-value
  • Water vapor permeability of 0.02 perm
  • Density of 2 lb/cu ft
  • Dimensions: 24 in. width x 4 ft. length x 2 in. thickness
  • Compliant with ASTM D1622, C518, D2126, E84, and E96 standards
Catalog No. 50-278-2449 Supplier Grainger Supplier No. 19NE93
These insulation boards are manufactured from polyisocyanurate (PIR) foam and feature FSK facing. Designed for high-performance thermal insulation, these boards are lightweight, rigid, and offer high-compression strength, making them a robust alternative to traditional insulation blankets and wraps. This insulation is suitable for applications requiring a temperature range of -297°F to +297°F.
  • Closed cell foam construction
  • Thermal resistance of 10.5 R-value
  • Water vapor permeability of 0.02 perm
  • Density of 2 lb/cu ft
  • Dimensions: 24 in. width x 4 ft. length x 2 in. thickness
  • Compliant with ASTM D1622, C518, D2126, E84, and E96 standards