Learn More
Cole-Parmer Megger MIT300-EN Insulation Tester 250V 500V Incl:Test Lead Set Crocodile Clips
Catalog No. 50631600 Shop All Cole-Parmer Products
Encompass
MIT300 series insulation and continuity testers
FEATURES
  • Tough rubber armored case
  • 250V, 500V and 1000V
  • Extremely easy to use
  • Self contained protective cover
  • Live circuit warning
  • Weatherproof to IP54
  • Intelligent safety system for protection
  • USB interface
Catalog No. 50-631-600 Supplier Cole-Parmer Supplier No. 2002250
MIT300 series insulation and continuity testers
FEATURES
  • Tough rubber armored case
  • 250V, 500V and 1000V
  • Extremely easy to use
  • Self contained protective cover
  • Live circuit warning
  • Weatherproof to IP54
  • Intelligent safety system for protection
  • USB interface