Learn More
Grainger ROCKWOOL Rockboard 40 Insulation Board, Stone Wool, 2 in, 24 in x 4 ft
Catalog No. 502779134
Encompass_Preferred
Rockboard 40 is a multipurpose stone wool insulation board designed for thermal and acoustic applications in walls, ceilings, and floors. This high-performance board provides effective noise reduction and fire protection in various settings, including mechanical rooms, utility rooms, and sound studios.
  • Noncombustible and fire resistant with a 0 flame spread index and 0 smoke developed index.
  • Maintains dimensional stability and will not slump, shrink, or expand with temperature changes.
  • Excellent sound-dampening characteristics across a wide range of frequencies.
  • Water and moisture repellent material that does not rot, corrode, or support mold growth.
  • Passes tests for corrosiveness to steel and stress corrosion cracking on austenitic stainless steel.
  • R-value of 4.2 hr.ft2.F/Btu per "
  • Hot surface performance rated up to 1,200 °F.
Catalog No. 50-277-9134 Supplier Grainger Supplier No. 19NE78
Rockboard 40 is a multipurpose stone wool insulation board designed for thermal and acoustic applications in walls, ceilings, and floors. This high-performance board provides effective noise reduction and fire protection in various settings, including mechanical rooms, utility rooms, and sound studios.
  • Noncombustible and fire resistant with a 0 flame spread index and 0 smoke developed index.
  • Maintains dimensional stability and will not slump, shrink, or expand with temperature changes.
  • Excellent sound-dampening characteristics across a wide range of frequencies.
  • Water and moisture repellent material that does not rot, corrode, or support mold growth.
  • Passes tests for corrosiveness to steel and stress corrosion cracking on austenitic stainless steel.
  • R-value of 4.2 hr.ft2.F/Btu per "
  • Hot surface performance rated up to 1,200 °F.