Learn More
Grainger Roxul High Temperature Insulation, Mineral Wool, 1-1/2 in. Thick, 7/Pk.
Catalog No. 502779192
Encompass_Preferred
These mineral wool insulation boards offer a lightweight, rigid solution for high-temperature applications. Designed for strength and durability, these boards are easier to cut than traditional insulation blankets and wraps while providing high-compression resistance.
  • Provides an R-value of 6
  • Constructed from mineral wool material
  • Features a thickness of 1-1/2 in. with an overall length of 4 ft. and a width of 24 in.
  • Maintains an operating temperature range of 0°F to 1,200°F
  • Features an unfaced design
  • Meets standards including ASTM C612 Type 1VB, C665, and C795 stainless steel
  • Chemically inert and does not absorb moisture
Catalog No. 50-277-9192 Supplier Grainger Supplier No. 19NE77
These mineral wool insulation boards offer a lightweight, rigid solution for high-temperature applications. Designed for strength and durability, these boards are easier to cut than traditional insulation blankets and wraps while providing high-compression resistance.
  • Provides an R-value of 6
  • Constructed from mineral wool material
  • Features a thickness of 1-1/2 in. with an overall length of 4 ft. and a width of 24 in.
  • Maintains an operating temperature range of 0°F to 1,200°F
  • Features an unfaced design
  • Meets standards including ASTM C612 Type 1VB, C665, and C795 stainless steel
  • Chemically inert and does not absorb moisture