Learn More
Grainger ROCKWOOL Mineral Wool High Temperature Insulation Board, 24 in. x 48 in., 0°F to 1200°F, FSK Facing
Catalog No. 502780022
Encompass_Preferred
This mineral wool high temperature insulation board is designed for thermal and acoustic applications in building construction, providing effective fire protection. Engineered to maintain its structure under extreme conditions, this insulation board serves as a reliable barrier for mechanical, utility, and acoustic environments.
  • Provides high-performance thermal resistance with an approximate R-value of 4 per "
  • Exhibits excellent acoustic dampening properties suitable for noise buffering.
  • Fire resistant and non-combustible to prevent flame spread.
  • Maintains dimensional integrity and will not slump, shrink, or expand with temperature.
  • Operates effectively within a temperature range of 0°F to 1,200°F.
  • Passes tests for corrosiveness to steel and stress corrosion cracking of austenitic stainless steel.
  • Compliant with ASTM C612 Type IVA and IVB standards.
Catalog No. 50-278-0022 Supplier Grainger Supplier No. 19NE81
This mineral wool high temperature insulation board is designed for thermal and acoustic applications in building construction, providing effective fire protection. Engineered to maintain its structure under extreme conditions, this insulation board serves as a reliable barrier for mechanical, utility, and acoustic environments.
  • Provides high-performance thermal resistance with an approximate R-value of 4 per "
  • Exhibits excellent acoustic dampening properties suitable for noise buffering.
  • Fire resistant and non-combustible to prevent flame spread.
  • Maintains dimensional integrity and will not slump, shrink, or expand with temperature.
  • Operates effectively within a temperature range of 0°F to 1,200°F.
  • Passes tests for corrosiveness to steel and stress corrosion cracking of austenitic stainless steel.
  • Compliant with ASTM C612 Type IVA and IVB standards.