Learn More
Waters Corp Insulator, AutoSpec Inner Source
Catalog No. 50793192 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is the insulator used between the Ion chamber and the mounting plate
Catalog No. 50-793-192 Supplier Waters Corp Supplier No. M702762AD1S
This is the insulator used between the Ion chamber and the mounting plate