Learn More
Grainger 3M Scotchlok Self-Stripping Electrical T-Tap Connector, 14-18 AWG, 50/Pk.
Catalog No. 502794654
Encompass_Preferred
Scotchlok™ self-stripping electrical T-tap connectors are designed to tap into a wire in mid-span without the need for cutting or stripping the insulation. These connectors create a reusable quick-connect wiring termination by mating with a 0.250 in. male tab. The construction features a flame-retardant nylon insulator and tin-plated brass element, suitable for use in appliances, light fixtures, and other OEM electrical applications.
  • Self-stripping design eliminates the need for wire cutting or stripping
  • Includes a hinged lid for secure termination
  • Fully insulated for safety
  • Constructed from flame-retardant nylon
  • Compatible with 14 AWG to 18 AWG copper conductors
  • Temperature rating of 221 °F
  • Maximum voltage rating of 600 V
  • Certified to CSA standards and UL listed
Catalog No. 50-279-4654 Supplier Grainger Supplier No. 1A074
Scotchlok™ self-stripping electrical T-tap connectors are designed to tap into a wire in mid-span without the need for cutting or stripping the insulation. These connectors create a reusable quick-connect wiring termination by mating with a 0.250 in. male tab. The construction features a flame-retardant nylon insulator and tin-plated brass element, suitable for use in appliances, light fixtures, and other OEM electrical applications.
  • Self-stripping design eliminates the need for wire cutting or stripping
  • Includes a hinged lid for secure termination
  • Fully insulated for safety
  • Constructed from flame-retardant nylon
  • Compatible with 14 AWG to 18 AWG copper conductors
  • Temperature rating of 221 °F
  • Maximum voltage rating of 600 V
  • Certified to CSA standards and UL listed