Learn More
Carbosynth LLC. Mouse monoclonal Insulin antibody
SDP
Supplier:  Carbosynth LLC. 10RI134E1MG
Encompass_Preferred
Insulin causes the cells in the liver, muscle, and fat tissue to take up glucose from blood and convert it to glycogen that can be stored in the liver and muscles. Insulin also prevents the utilization of fat as an energy source. In absence of insulin or in conditions where insulin is low glucose is not taken up by body cells, and the body begins to use fat as an energy source
Catalog No. 50-125-2756