Learn More
CPC Scientific H-Glu-Ala-Leu-Tyr-Leu-Val-Cys-Gly-Glu-Arg-Gly-OH (ammonium salt) 1MG
SDP
Supplier:  CPC Scientific PANP007A
Encompass
This peptide is the 13-23 fragment of the insulin B chain. This sequence represents an important T cell epitope for islet-infiltrating T cells in NOD mice.ONE-LETTER SEQUENCE: EALYLVCGERGMOLECULAR FORMULA: C52H84N14O17S1MOLECULAR WEIGHT:1209.38STORAGE CONDITIONS: -20 5CRESEARCH AREA: GastrointestinalREFERENCES: Lee, K. et al. Nat. Immunol. 2, 501 (2001)
Catalog No. 50-211-0083