Learn More
Eurofins Discoverx PathHunter Insulin Bioassay Kit, 10-Plate. (93-0466Y3-00008)
Catalog No. 502000586
Encompass
The PathHunter Insulin Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses insulin based activation of INSRb RTK receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged INSRb and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0586 Supplier Eurofins Discoverx Supplier No. 930466Y300008
Add to Cart
Add to Cart
Request a Quote
The PathHunter Insulin Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses insulin based activation of INSRb RTK receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged INSRb and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.