Learn More
Medchemexpress LLC Insulin protein, bovine (P. pastoris) | ≥95.0% | C254H377N65O75S6 | 100MG
SDP
Catalog No. 5000174982
Encompass_Preferred
Recombinant bovine insulin produced in Pichia pastoris and supplied lyophilized for research use. The protein is native-sequence insulin with an approximate molecular weight of 5,733 Da and purity ≥95%, suitable for biochemical assays, receptor binding studies, and cell culture applications. It has low endotoxin levels (<0.1 EU/μg) and is stable when stored frozen; reconstitute per guidance for experimental use.
  • Native-sequence bovine insulin for biological studies.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.1 EU/μg) suitable for sensitive assays.
  • Lyophilized formulation for improved stability and transport.
  • Stable when stored at -20 °C; aliquot and freeze for long-term storage.
  • Reconstitute at ≥1 mg/mL in ddH2O for experimental use.
Catalog No. 50-001-74982 Supplier Medchemexpress LLC Supplier No. HYP701240100MG
Recombinant bovine insulin produced in Pichia pastoris and supplied lyophilized for research use. The protein is native-sequence insulin with an approximate molecular weight of 5,733 Da and purity ≥95%, suitable for biochemical assays, receptor binding studies, and cell culture applications. It has low endotoxin levels (<0.1 EU/μg) and is stable when stored frozen; reconstitute per guidance for experimental use.
  • Native-sequence bovine insulin for biological studies.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.1 EU/μg) suitable for sensitive assays.
  • Lyophilized formulation for improved stability and transport.
  • Stable when stored at -20 °C; aliquot and freeze for long-term storage.
  • Reconstitute at ≥1 mg/mL in ddH2O for experimental use.