Learn More
Medchemexpress LLC HY-P1156 10mg Medchemexpress, Insulin(cattle) CAS:11070-73-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP115610MG
Encompass_Preferred
Medchemexpress, HY-P1156 10mg Insulin(cattle) CAS:11070-73-8 Insulin cattle (Insulin from bovine pancreas) is a two-chain polypeptide hormone produced in vivo in the pancreatic β cells. Insulin cattle has often been used as growth supplement in culturing cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4156